


Silver sulphate ≥98.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # BAKR3443-04
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:125 g
- Seq:3445
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:= 98.0 %
- For Laboratory,Research,or Manufacturing Use:= 0.02 %
- ACS - Assay (Ag₂SO₄):≥98.0 %
- ACS - Insoluble Matter and Silver Chloride:≤0.02 %
- ACS - Nitrate (NO₃):≤0.001 %
- ACS - Substances Not Precipitated by HCl:≤0.03 %
- ACS - Iron (Fe):≤0.001 %
Specification Test Results
About this item
:
:311.8 g/mol
Merck Index:13,08603
Boiling Pt:1085 °C (1013 hPa)
Melting Pt:655 °C
Density:5,45 g/cm³ (20 °C)
EINECS:233-653-7
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:10294-26-5
Formula:Ag₂SO₄
MDL Number:MFCD00003407