
2-Butoxyethanol ≥99% (by GC), J.T.Baker®
Catalog # BAKRD648-07
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99 %
- Assay (CH₃(CH₂)₃OCH₂CH₂OH) (by GC):≥99 %
- Color (APHA):≤10
Specification Test Results
About this item
:
:118.18 g/mol
Merck Index:13,01558
Boiling Pt:171 °C (1013 hPa)
Melting Pt:–70 °C
Density:0,902 g/cm³ (20 °C)
Flash Pt:61 °C
Storage Temperature:Ambient
EINECS:203-905-0
UN:2810
ADR:6.1,III
:GHS06
CAS Number:111-76-2
Formula:CH₃(CH₂)₃OCH₂CH₂OH
MDL Number:MFCD00002884