Order Entry
Singapore
ContactUsLinkComponent
 
Ethylenediamine ≥98.0%, BAKER ANALYZED®, J.T.Baker®
 :  Chemicals
 
undefined
Ethylenediamine ≥98.0%, BAKER ANALYZED®, J.T.Baker®
 :  Chemicals
 

 

  • Environmentally Preferable:

 

 

  • For Laboratory,Research,or Manufacturing Use:
    = 98.0 %
  • Assay (NH₂CH₂CH₂NH₂):
    ≥98.0 %
  • Density (g/mL) at 25°C:
    0.890 - 0.903
  • Heavy Metals (as Pb):
    ≤0.001 %