Cylinder bench clamp, Talon®
Catalog # TROE971056
Supplier: TROEMNER INSTRUMENTS
Specifications
- Description:Bench clamp with strap
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Cast aluminium clamp.
- Mounts cylinders to bench or table top
These clamps feature a 25 mm wide, 1372 mm long nylon strap with nickel plated, non slip spring catch and buckle for fast, easy adjustment.