Volumetric flasks, class A, glass, clear, NS glass stopper, PYREX®
Supplier: Corning
Certificates
About this item
Made of PYREX® borosilicate glass.
- Excellent chemical resistance
- Excellent temperature resistance
Reusable volumetric flasks with glass stoppers. This flasks have an upper bulb with the one-tenth of flask rated capacity for the mixing of liquids or powders when shaken.
Certifications: ASTM E-694, E-542, E-288.