VWR®, Embedding compound, water- soluble
Catalog # VWRU95057-838
Supplier: VWR Collection
CAS Number:
Specifications
- Pk:1
- Description:Section compound Cl, premium, frozen
Specifications
About this item
Frozen section compound is a fast-acting, water-soluble embedding compound designed for use in frozen sectioning. The compound bonds and encapsulates tissue specimens to the object holder for cryosectioning. At a working temperature of –20 °C, it provides optimal sectioning consistency with minimal section curling.
- Highly recommended for surgical pathology laboratories
- Available clear or in a light blue format
- Each bottle features an extended length tip for increased accuracy in dispensing
- Size: 118 ml