Order Entry
Singapore
ContactUsLinkComponent
Surface disinfectant solution, Sporicidin®
Surface disinfectant solution, Sporicidin®
Catalog # CONTRE-1284F
Supplier:  CONTEC
Surface disinfectant solution, Sporicidin®
Catalog # CONTRE-1284F
Supplier:  CONTEC
Specifications
  • Description:
    Sporicidin surface disinfectant, 3,8 l container
  • Packed:
    4× 3,8 litres (1 gallon)
  • Pk:
    4
Specifications
About this item
Sporicidin Disinfectant Solution is an EPA registered intermediate level disinfectant for use on environmental surfaces in fitness industries, medical offices, pharmaceutical cleanrooms, compounding pharmacies, prisons, and animal labs where broad-spectrum kill claims and surface compatibility are needed.
  • Broad spectrum kill claim
  • Six month residual activity against odour causing bacteria
  • Excellent health and safety profile
  • Mild alkaline pH
  • No intermediate level disinfectant rotation required if proper protocol followed
Sporicidin is based on buffered phenol, sodium phenate, and surfactant blends which help to provide both cleaning and disinfecting activity in one product.