VWR® Traceable®, Digital refrigerator and freezer thermometers
Catalog # VWRU61161-364
Supplier: VWR Collection
Specifications
- Range:–50…+70 °C
- Resolution:1 °C
- Weight:113 g
- W×D×H:70×108×19 mm
- Accuracy:±1,0 °C
- Pk:1
- Probe:Bottle probe
Specifications
About this item
These thermometers accurately monitor temperatures in freezers, water baths, heating blocks, incubators, and refrigerators with this enclosed temperature-buffered sensor.
- Triple display simultaneously shows min/max and current temperatures, °F/°C switchable
- Bottle insulates sensor from rapid temperature changes when refrigerator door is opened
- Ten-foot micro-cable permits refrigerator doors to close on it
- High and low settable alarms
Non toxic glycol bottle probe.
Certifications: Traceable to NIST. Calibration compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001. Meets current CDC requirements and additional recommendations for vaccine storage and monitoring. Can be used in environments requiring 21 CFR 11 compliance.
Delivery: Supplied with Traceable® certificate, flip-open stand, wall mounting, Velcro®, and magnetic strips.