Precision weights, OIML, class E2
5 mg, Individual weight, Class E2
Catalog # TROE7534-E2W
Supplier: TROEMNER INSTRUMENTS
Jump to:
Product Details & Documents
These individual milligram weights are manufactured from the finest stainless steel. Available in flat or wire construction.
: Accredited by National Voluntary Laboratory Accreditation Programme under Lab Code 105013-0 and Statement of Accuracy.
: Supplied in attractive, durable, high quality polycarbonate cases with a compartment for each weight within the case. There is also a compartment for a brush, forceps, and a USB flash drive which contains the electronic certificate.
Specifications
- Weight:5 mg
- Type:Individual weight, Class E2