Order Entry
Singapore
ContactUsLinkComponent
VWR®, Spatula
Catalog # VWRU82027-534
Supplier:  VWR Collection
undefined
VWR®, Spatula
Catalog # VWRU82027-534
Supplier:  VWR Collection
Specifications
  • Length:
    178 mm
  • Pk:
    1
Specifications
About this item
Stainless steel, PTFE coated. This spatula features one rounded end and one flat end, with PTFE coating throughout.
Can be used to transfer corrosive or sticky materials without scratching or damaging container surfaces.