Measuring cylinders, tall form, class A, borosilicate glass, PYREX®
Supplier: Corning
Certificates
About this item
Borosilicate glass, class A.
- Calibrated to deliver (TD) in accordance with ASTM E-1272 and ASTM E-542
- Cylinders are individually marked with serial number
- White graduation
- 10 ml cylinder has a funnel top
Delivery: Cylinders are supplied with bumper guard for 25 to 2000 ml and also with a certificate of identification and capacity, traceable to NIST.