Mixing measuring cylinders, tall form, PYREX®
: Corning
Borosilicate glass, without stoppers.
- Calibrated 'to contain' (TC)
- Equipped with a 24/40 ST joint
- Single metric scale
- Graduations are in durable white enamel
Borosilicate glass, without stoppers.