Volumetric flasks, class A, low actinic, PYREX® EZ Access™
Supplier: Corning
Certificates
About this item
PYREX® glass, low actinic, with stopper.
- Graduation line is sharp and permanent
- Low actinic glass provides protection for materials sensitive to light
- Chemical and thermal resistance
Ideal for microchemical work. Features wide mouth facilitates pipette filling and mixing.
The flasks are supplied with larger interchangeable glass stoppers.