Separating funnels, globe shaped, PYREX®
1000 ml
Supplier: Corning
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, with glass stopper.
These separatory funnels are made from molded blanks which are tapered toward the stopcock to facilitate the separation of liquids. The sturdy design makes it suitable for rack work.
Documents
Certificates
Search by Lot NumberSpecifications
- Capacity:1000 ml
- Seq:04