Sulphuric acid 0.5 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
BAKR5640-02
: Avantor
:
- Pack type:PE bottle, narrow mouth
- Pk:1 L
- Seq:5037
- For Laboratory,Research,or Manufacturing Use:0.4995 - 0.5005
- Normality:0.4995 - 0.5005
- Chloride (Cl):≤1 ppm
- Phosphate (PO₄):≤1 ppm
- Heavy Metals (as Pb):≤0.3 ppm
- Trace Impurities - Iron (Fe):≤0.5 ppm
:
:98.08 g/mol
:1,01 g/cm³ (20 °C)
:Ambient
:2796
:8,II
:Management of Change (MOC) category = NI
:231-639-5
:7664-93-9
:H₂SO₄
:MFCD00064589