


Nitric acid 65% Huey, BAKER ANALYZED® for corrosion testing, J.T.Baker®
Catalog # BAKR9597-33
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, clear, PVC-coated
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:64.8 - 65.2 %
- Assay (as HNO₃) (by acidimetry):64.8 - 65.2 %
- Appearance:Passes Test
- Color (APHA):≤ 10
- Residue after Ignition:≤ 0.0005 %
- Chloride (Cl):≤ 0.05 ppm
- Phosphate (PO₄):≤ 0.20 ppm
- Sulfate (SO₄):≤ 0.5 ppm
- Trace Impurities - Arsenic (As):≤ 0.005 ppm
- Trace Impurities - Chromium (Cr):≤ 0.100 ppm
- Trace Impurities - Copper (Cu):≤ 0.050 ppm
- Heavy Metals (as Pb):≤ 0.1 ppm
- Trace Impurities - Iron (Fe):≤ 0.1 ppm
- Trace Impurities - Nickel (Ni):≤ 0.050 ppm
- Fluoride (F):≤ 1.000 ppm
Specification Test Results
About this item
:
:63.01
Merck Index:13,06608
Boiling Pt:120,5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1,4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
:Management of Change (MOC) category = NI
:GHS03
:GHS05
:GHS07
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349