


Glutaraldehyde 25% in water, Practical grade, J.T.Baker®
Catalog # BAKRM752-07
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Identification:Passes Test
Specification Test Results
About this item
:
:100.12 g/mol
Boiling Pt:101 °C (1013 hPa)
Melting Pt:–6 °C
Density:1,06 g/cm³ (20 °C)
Flash Pt:>71 °C
Storage Temperature:Refrigerator
EINECS:203-856-5
UN:3265
ADR:8,III
:GHS05
:GHS08
:GHS09
CAS Number:111-30-8
Formula:C₅H₈O₂
MDL Number:MFCD00007025