Specific gravity bottles, Gay-Lussac, PYREX®
CORN1622-2
: Corning
- Volume (nom.):2 ml
- Pk:1
PYREX® borosilicate glass, cylindrical.
- Reusable
- With hollow glass stopper (NS 7/15)
These specific gravity bottles are unadjusted for calibration in the laboratory.