Order Entry
Singapore
Orders LinkContactUsLinkComponent
Roller bottle, PYREX®

Roller bottle, PYREX®

840 cm², 110×285 mm, 168 - 252 ml
Catalog # CORN1400-285
Supplier:   Corning
Roller bottle, PYREX®
Catalog # CORN1400-285
Supplier:  Corning
Roller bottle, PYREX®
Roller bottle, PYREX®

840 cm², 110×285 mm, 168 - 252 ml

Price
$440.00
Log in for availability
1 * 2 items
Product Details & Documents

PYREX®, borosilicate glass.

  • Optical clarity and mechanical strength
  • Withstand wet or dry sterilisation

 : Supplied with 38 mm deep skirted GPI 38-430 threaded phenolic screw cap with rubber liner.

 : *The product(s) on this page are outside the scope of the EU Directive 2019/904

Specifications
  • Growth area:
    840 cm²
  • Recommended working volume:
    168 - 252 ml
  • Ø ext.×H:
    110×285 mm