Order Entry
Singapore
ContactUsLinkComponent
VWR®, Aprons, PE
VWR®, Aprons, PE
Catalog # VWRU10826-007
Supplier:  VWR Collection
undefined
VWR®, Aprons, PE
Catalog # VWRU10826-007
Supplier:  VWR Collection

Specifications

  • Colour:
    White
  • Pk:
    100
  • W×L:
    711×1168 mm
  • Thickness:
    0,05 mm

Specifications

About this item

This PE apron helps to keep clothing clean and dry.

  • Disposable
  • Medium-weight fabric
  • One-piece bib design just slips over the head and ties in back
Packaging: 100/dispenser box.