VWR®, Culture tube, disposable, with screw cap finish
Supplier: VWR Collection
Certificates
About this item
Borosilicate glass 3.3, disposable.
Tubes have a marking spot and reference mark for orientation during use, and a threaded neck to accept screw cap.
Ordering information: Caps must be ordered separately.