Order Entry
Singapore
Orders LinkContactUsLinkComponent
VWR®, Tissue Grinders, PTFE
Supplier:   VWR Collection
VWR®, Tissue Grinders, PTFE
Supplier:  VWR Collection
Certificates
VWR®, Tissue Grinders, PTFE
VWR®, Tissue Grinders, PTFE
4 Options Available Below
Product Details & Documents
PTFE, stainless steel and precision borosilicate glass. Designed to control the reduction of particle size and homogenisation of variety of substances (biological material). Shearing forces will be generated by the movement of a rotating plunger in a precision bore tube.
  • Chemically inert and leak free
  • Non-contaminating
  • Usable up to 280 °C
 : Approved by FDA and USP Class VI.
Product Family Options
Glass vessel
Product Information
  • Capacity
    Ø ext.×length
    Total length
    Availability
    Price
  • 2 ml
    8×120 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $193.00
    $193.00
    1 * 1 items
       $193.00
  • 5 ml
    12×135 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $238.00
    $238.00
    1 * 1 items
       $238.00
Plunger
Product Information
  • Capacity
    Ø ext.×length
    Total length
    Availability
    Price
  • 2 ml
    230 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $83.80
    $83.80
    1 * 1 items
       $83.80
  • 5 ml
    235 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $86.70
    $86.70
    1 * 1 items
       $86.70