VWR®, Tubing, Silicone
Platinum-cured silicone tubing is flexible, durable, low volatile grade, and translucent. This tubing is ideal for a variety of transfer applications such as pharmaceutical, laboratory, food and beverage, and bioprocess manufacturing. It features an ultra-smooth interior surface that provides uniform fluid flow and resistance to bacteria build-up.
- Can be sterilised by autoclaving and gamma-irradiation
- Imparts no taste or odour to critical stream
: Meets USP Class VI; USP 661 (Physicochemical Tests); ISO10993, EP 3.1.9, 3-A standards and FDA CFR 177.2600.
: Available in single polyethylene pack 50' coil lengths with quality control and lot traceability.
- Catalog No:
- VWRU89068-474
- VWRU89068-454
- VWRU89068-478
- VWRU89068-482
- VWRU89068-490
- VWRU89068-456
- VWRU89068-488
- VWRU89068-468
- VWRU89068-436
- VWRU89068-432
- Durometer hardness:
- Shore A, 50
- Shore A, 50
- Shore A, 50
- Shore A, 50
- Shore A, 50
- Shore A, 50
- Shore A, 50
- Shore A, 50
- Shore A, 50
- Shore A, 50
- Max. temperature:
- 204 °C
- 204 °C
- 204 °C
- 204 °C
- 204 °C
- 204 °C
- 204 °C
- 204 °C
- 204 °C
- 204 °C
- Min. temperature:
- −73 °C
- −73 °C
- −73 °C
- −73 °C
- −73 °C
- −73 °C
- −73 °C
- −73 °C
- −73 °C
- −73 °C