Standard system for recycling aerosol cans, Aerosolv®
Catalog # JUSU28202
Supplier: JUSTRITE MANUFACTURING
Specifications
- Description:Aerosolv® can disposal system
- Pk:1
Specifications
About this item
This standard Aerosolv® system includes puncturing unit with plastic sleeve, colour changing coalescing/carbon filter cartridge, anti-static wire and goggles to enhance safety. Made of low maintenance aircraft aluminum and threads into standard 2" bung on 110 and 200 litre drums.
- Filter removes liquids and adsorbs odours
- Cans are recyclable as scrap metal
- All moving parts are 308 stainless steel
- Carbide tipped piercing pin shows no signs of wear after puncturing 10000 cans
Certifications: Complies with EPA 40 CFR 261.23(a)(6).