Culture flasks, low form, PYREX®
Catalog # CORN4422-2XL
Supplier: Corning
Specifications
- Ext.Ø:254 mm
- Capacity:2500 ml
- Height:197 mm
- Neck Ø ext.:38 mm
- Pk:1
Specifications
About this item
PYREX® borosilicate glass.
- Very good temperature resistance
- Very good chemical resistance
These low form culture flasks are ideal for culturing organisms that require a large surface area-to-volume ratio, and in serum production. These flask have a heavy beaded rim to protect against chipping and include a large marking spot allows for accurate labeling and easy identification.
Caution: Do not use this flask on a heat source smaller than the base of the flask. Uneven heating of the flask could cause breakage.