Stopcocks, plain bore, PYREX® Rotaflo®
: Corning
PYREX®, borosilicate glass.
- Fine flow adjustment
- Lubrication is not required
- Fast assembly
These high performance Rotaflo® stopcocks can be adjusted with bore openings and have sidearms. Only chemically resistant PTFE and PYREX® borosilicate glass are in contact with liquids and gase. Seal is visible when open or closed.