Order Entry
Singapore
ContactUsLinkComponent
Tissue grinders, Tenbroeck type, PYREX®
Tissue grinders, Tenbroeck type, PYREX®
Catalog # CORN7726-L
Supplier:  Corning
undefined
Tissue grinders, Tenbroeck type, PYREX®
Catalog # CORN7726-L
Supplier:  Corning

Specifications

  • Description:
    Homogeniser, complete, size - L
  • Total length:
    340 mm
  • Pk:
    1
  • Ø ext.×length:
    20×335 mm

Specifications

About this item

These homogenisers are designed for grinding small samples for pathological and biochemical work. The catch basin bulb has a 55 mm outer diameter.

Can be used manually but is adaptable for use with a slow speed stirrer by means of a rubber stopper and a connecting rod.

Certifications: Made in accordance with Federal Specification DD-T-363a.
Caution: If used with a motor-driven device, the attachment must contain some form of friction release clutch to prevent damage to the apparatus. The user should wear a protective glove of sufficient thickness to prevent injury in the event of failure of the glass components.