Order Entry
Singapore
Orders LinkContactUsLinkComponent
Settlometer, Mallory, PYREX®

Settlometer, Mallory, PYREX®

Mallory direct reading settlometer, 2000 ml
Catalog # CORN7250-2L
Supplier:   Corning
Settlometer, Mallory, PYREX®
Catalog # CORN7250-2L
Supplier:  Corning
Settlometer, Mallory, PYREX®

Mallory direct reading settlometer, 2000 ml

Price
$616.00
Log in for availability
1 * 1 items
Product Details & Documents

Used with oxidised sludge process for treatment of sewage and other wastes. These cylinders are graduated in cc per litre and in hundredths of a foot.

Int.Ø×H: 130×190,5 mm.

Documents

Specifications
  • Description:
    Mallory direct reading settlometer, 2000 ml