Mixing measuring cylinders, tall form, class A, borosilicate glass, PYREX®
Supplier: Corning
Certificates
About this item
PYREX® borosilicate glass, class A.
- Single metric scale cylinders are individually serialised
- Calibrated to contain (TC) in accordance with E-1272 and ASTM E-542
- White enamel graduations
PYREX® Serialised/Certified Volumetric Glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Graduation markings are in durable white enamel. Single metric scale. With [ST] PYREX ®stopper.
Delivery: Supplied with a Corning Certificate of Identification and Capacity, traceable to NIST standards.