Burettes, serialized, class A, white graduations, PYREX®
: Corning
PYREX® borosilicate glass, class A.
- Durable white graduations, sharply defined
- Easy to read
These precision bore tubing is used to assure accuracy between any two points. Each burette is individually serialised and supplied with a certificate of identification and capacity, traceable to NIST standards.
Capacity tolerance is established by ASTM E-542 and ASTM E-287.
PYREX® Serialised/Certified Volumetric Glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Durable coloured graduations are sharply defined and easy-to-read. Carefully ground [ST] stopcock gives precise control.
: Burettes are supplied with bore stopcock plug and dust cover.