Burettes, straight bore PTFE stopcock, PYREX®
: Corning
Borosilicate glass.
- Microfinish of the barrel provides a precision fit
- Durable coloured markings
These burettes are suitable for use in school and institutional laboratories where the accuracy of class A models is not required.
The tips are carefully drawn for accuracy to insure proper drainage rate.
: Burettes are supplied with a 2 mm PTFE stopcock plug and dust cover.