Burettes, PTFE stopcock, serialised, PYREX®
Supplier: Corning
Certificates
About this item
Borosilicate glass, class A.
- Individually serialised
- Coloured markings with fine sharp lines
- Large easy to read numbers
- Smooth micro-finish stopcock assures leak-resistant performance
Burette tips are carefully drawn from precision bore tubing to ensure proper drainage rates.
Designed as per ASTM E287 and calibrated in accordance with ASTM E542 and D-511 requirements. Traceable to NIST standards.
Certifications: PYREX® Serialised/Certified Volumetric Glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Delivery: Burettes are supplied with 2 mm bore PTFE stopcock plug, a dust cover and certificate of identification and capacity.