Digital measuring devices, PHYSICS 300
Supplier: Ludwig Schneider GmbH & CO. KG
Certificates
About this item
PHYSICS 300 provides a wide range of measuring units and sensor technologies with data logger function. It is suitable for measuring temperature, air humidity, air flow, pressure, flow rate, electrical values etc. With its data logger functions, it provides precise measurements in meteorology, water analystics, chemistry, biotechnology, industry, research and development etc. Three measuring points can be covered simultaneously.
- Compact shape and ergonomic design
- Intelligent probe display with probe specific functions
- With three input probes for 65 measuring units
- Language selection - English, German or French
- With graphic display, 128×64 pixel, 8 lines with 2 white LEDs
- Measuring functions inlcude measuring value, zero-setting, max./min. value documentation, 2-step adjustment
- Used in test functions like segment monitoring, range monitoring, sensor error check and battery check
Ordering information: Available with work and DAkkS certificate. ABS case and all other accessories can be ordered separately.