Mini bioreactor centrifuge tubes, Corning®
Catalog # CORN431720
Supplier: Corning
Specifications
- Closure type:Screw cap
- Ø×L:29,1 × 114,4 mm
- Type:Conical bottom
- Capacity:50 ml
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Pk:300
- RCF max.:12000×g
Specifications
About this item
PP, translucent, with orange PP screw cap.
- Large marking spot enables clear identification and experimental parameters
- Easy-to-read graduations
- Non pyrogenic
- Free from RNase and DNase
- Sterile
Ideal for high throughput process optimisation for suspension cell culture, the Mini Bioreactor consists of a centrifuge tube with vented cap. Tube applications include cell line developement, clone selection, media optimisation, and recombinant protein development. Screw cap features four vent holes and a 0,22 µm hydrophobic membrane to allow for gas exchange.