Repetitive pipette, Multipette® E3 / E3x
Supplier: EPPENDORF
Certificates
About this item
This pipette enables precise pipetting from 1 μl to 50 ml and makes long pipetting series faster and easier. The aspirated liquid can be dispensed up to 100 times without a refill making the Multipette® the ideal tool for filling plates or large series of tubes (e.g., nucleic acid purification kits).
- To be used with Eppendorf Combitips advanced®: Automatic tip recognition for ease of use
- Defined dispensing speed increases the reproducibility of results; eight different speed levels allow the optimum speed for each application
- One button tip ejection enables one handed operation
- RFID chip contains all relevant data regarding the pipette (serial no., dispensing vol., factory adjustment, and more)
- Enlarged colour display with optimised contrast and clear arrangement of all adjustable parameters is easy to read
- Software shows all selected parameters at one glance; proven intuitive software navigation
- Display in 9 different languages
Operation modes:
E3 model: Dispensing, automatic dispensing, pipetting
E3x model: Dispensing, automatic dispensing, pipetting, aspirating, titrating, sequential dispensing, combined aspiration and dispensing mode
* Works with Windows 7/8/Vista.
Delivery information: Supplied with Combitips advanced® assortment pack (one Combitip of each size).
Specifications
- Catalog No:
- EPPE4987000.010
- EPPE4987000.029
- EPPE4987000.371
- EPPE4987000.380
- Environmentally Preferable:
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Environmental certification:
- Renewable Energy Certificate, ISO 14001, ACT Label
- Renewable Energy Certificate, ISO 14001, ACT Label
- Renewable Energy Certificate, ISO 14001, ACT Label
- Renewable Energy Certificate, ISO 14001, ACT Label