Anti-POMC Mouse Monoclonal Antibody [clone: B31.15]
Catalog # ABNOMAB0905
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Proopiomelanocortin
- Antigen symbol:POMC
- Clonality:Monoclonal
- Clone:B31.15
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:5443
- Antigen synonyms:OTTHUMP00000119991|adrenoCorticotropin/ beta-lipotropin/ alpha-melanocyte stimulating hormone/ beta-melanocyte stimulating|NPP|beta-LPH|lipotropin beta|hormone/ beta-endorphin|alpha-melanocyte-stimulating hormone|adrenocorticotropic hormone|lipotropin gamm|POC|beta-melanocyte-stimulating hormone|gamma-MSH|alpha-MSH|beta-endorphin|CLIP|LPH|Precursor of MSH|adrenoCorticotropin|POMC|proopiomelanocortin|gamma-LPH|OTTHUMP00000200964|Corticotropin-like intermediary peptide|ACTH
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified human POMC.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native POMC.
Type: Primary
Antigen: POMC
Clonality: Monoclonal
Clone: B31.15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human