Anti-LPS Mouse Monoclonal Antibody [clone: B352M]
Catalog # ABNOMAB3948
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Lipopolysaccharide
- Antigen symbol:LPS
- Clonality:Monoclonal
- Clone:B352M
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Chlamydia
- Cross adsorption:No
- Format:Liquid
- Storage buffer:10 mM PBS, pH 7,2 (0,1% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native from Chlamydia trachomatis elementary bodies.
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against Chlamydia species LPS.
Recommended Dilutions: Recommend dilutions start at 5 ?g/ml.
Type: Primary
Antigen: LPS
Clonality: Monoclonal
Clone: B352M
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Chlamydia