Anti-CDKN2A Rat Monoclonal Antibody [clone: 5-C3-1]
ABNOMAB2417
: Abnova
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase inhibitor 2A
- Antigen symbol:CDKN2A
- Clonality:Monoclonal
- Clone:5-C3-1
- Conjugation:Unconjugated
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:12578
- Antigen synonyms:ARF, CDK4I, CDKN2, CMM2, INK4, INK4A, MLM, MTS-1, MTS1, P14, P14ARF, P16, P16-INK4A, P16INK4, P16INK4A, P19, P19ARF, TP16
- Amino acid number:54 to 75
- Storage buffer:Tris-glycine, 150 mM NaCl (0,05% sodium azide)
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 54-75 of mouse Cdkn2a.
- Size:100 μl
- Pk:100 µg
Rat monoclonal antibody raised against synthetic peptide of Cdkn2a.
Recommended Dilutions: Immunohistochemistry (Frozen sections): 5 ?g/ml; Western Blot: 1:500-1:1000
Type: Primary
Antigen: CDKN2A
Clonality: Monoclonal
Clone: 5-C3-1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse