Anti-DO-11.10 TCR Monoclonal Antibody [clone: KJ1-26.1]
ABNOMAB2424
: Abnova
- Antibody type:Primary
- Antigen symbol:DO-11.10 TCR
- Clonality:Monoclonal
- Clone:KJ1-26.1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Isotype:IgG2a
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (0,1% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified from DO-11.10 T cell hybridoma.
- Size:100 μl
- Pk:100 µl
Mouse monoclonal antibody raised against DO-11.10 TCR.
Type: Primary
Antigen: DO-11.10 TCR
Clonality: Monoclonal
Clone: KJ1-26.1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a
Reactivity: