Anti-CERK Rabbit Polyclonal Antibody
ABNOPAB12459
: Abnova
- Antibody type:Primary
- Antigen name:Ceramide kinase
- Antigen symbol:CERK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:64781
- Antigen synonyms:hCERK|Acylsphingosine kinase|Lipid kinase 4|LK4|KIAA1646
- Amino acid number:50 to 150
- Storage buffer:Tris-glycine, 150 mM NaCl (0,05% sodium azide)
- Storage temperature:Store at –20 °C or –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 50-150 of human CERK.
- Size:100 μl
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of CERK.
Recommended Dilutions: Western Blot: 2 ?g/ml
Type: Primary
Antigen: CERK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human