Anti-FAR2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB20854
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fatty Acyl CoA Reductase 2
- Antigen symbol:FAR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:55711
- Antigen synonyms:SDR10E2|MLSTD1
- Storage buffer:PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
- Sequence:FLGKVLMEKLFRTSPDLKVIYILVRPKAGQTLQQRVFQILDSKLFEKVKEVCPNVHEKIRAIYADLNQNDFAISKEDMQELLSCTNIIFHCAATVRFDD
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human FAR2.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against recombinant FAR2.
Recommended Dilutions: Immunohistochemistry: 1:20-1:50; Western Blot: 1:250-1:500
Type: Primary
Antigen: FAR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human