Anti-NID Mouse Monoclonal Antibody [clone: 331C9]
ABNOMAB3441
: Abnova
- Antibody type:Primary
- Antigen name:Nidogen
- Antigen symbol:NID
- Clonality:Monoclonal
- Clone:331C9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM
- Reactivity:Pig,Chicken,Human,Bovine
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Bovine nidogen.
- Size:100 μl
- Pk:100 µl
Mouse monoclonal antibody raised against Nidogen.
Recommended Dilutions: ELISA: 1:500-1:1000; Western Blot: 1:200-1:1000; Immunohistochemistry (Frozen sections): 1:100-1:300
Type: Primary
Antigen: NID
Clonality: Monoclonal
Clone: 331C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Bovine, Chicken, Human, Pig