Anti-NRCAM Mouse Monoclonal Antibody [clone: RNL-1]
ABNOMAB3561
: Abnova
- Antibody type:Primary
- Antigen name:Neuronal cell adhesion molecule
- Antigen symbol:NRCAM
- Clonality:Monoclonal
- Clone:RNL-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Form:Cell Culture Supernatant
- Gene ID:4897
- Antigen synonyms:Bravo|C030017F07Rik|mKIAA0343|C130076O07Rik
- Storage buffer:0,09% sodium azide
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Native purified NRCAM from NCI-H82 cell.
- Size:1 mL
- Pk:1 ml
Mouse monoclonal antibody raised against native NRCAM.
Recommended Dilutions: Immunohistochemistry: 1:10-1:25
Type: Primary
Antigen: NRCAM
Clonality: Monoclonal
Clone: RNL-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human