Anti-VCP Mouse Monoclonal Antibody [clone: Hs-14]
Catalog # ABNOMAB3824
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Valosin containing protein
- Antigen symbol:VCP
- Clonality:Monoclonal
- Clone:Hs-14
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:7415
- Antigen synonyms:ALS14|HEL-S-70|IBMPFD1|TERA|p97|HEL-220|IBMPFD
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Native purified human ACRV1.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native purified ACRV1.
Recommended Dilutions: Immunocytochemistry: 10 ?g/ml
Type: Primary
Antigen: VCP
Clonality: Monoclonal
Clone: Hs-14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse