Anti-5-mC Monoclonal Antibody [clone: 5MC-CD]
ABNOMAB6766
: Abnova
- Antibody type:Primary
- Antigen name:5-Methylcytosine
- Antigen symbol:5-mC
- Clonality:Monoclonal
- Clone:5MC-CD
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Antiserum
- Storage buffer:0,05% sodium azide
- Storage temperature:Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:5-Methylcytosine conjugated with BSA.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against 5-Methylcytosine.
Recommended Dilutions: Immunocytochemistry: 1:50-1:100; Western Blot: ~1:1000
Type: Primary
Antigen: 5-mC
Clonality: Monoclonal
Clone: 5MC-CD
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgM
Reactivity: