Anti-PTPRC Rat Monoclonal Antibody (Spectral Red®) [clone: C363.16A]
Catalog # ABNOMAB5734
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Monoclonal
- Clone:C363.16A
- Conjugation:Spectral Red®
- Flow cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:19264
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Storage buffer:PBS (1 mg/mL sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Cloned mouse Th2 cell lines.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rat monoclonal antibody raised against Ptprc exon B-dependent form.
Recommended Dilutions: Flow Cytometry: 0.2 ?g/10^6 cells
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: C363.16A
Conjugation: Spectral Red®
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse