Anti-FAT1 Mouse Monoclonal Antibody [clone: Fat-1]
Catalog # ABNOMAB7207
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:FAT atypical cadherin 1
- Antigen symbol:FAT1
- Clonality:Monoclonal
- Clone:Fat-1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgM
- Reactivity:Drosophila
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:2195
- Antigen synonyms:FAT|ME5|CDHF7|hFat1|CDHR8
- Storage buffer:PBS
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Cytoplasmic domain of drosphila FAT1.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against FAT1.
Type: Primary
Antigen: FAT1
Clonality: Monoclonal
Clone: Fat-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Drosophila