Anti-UXT Goat Polyclonal Antibody
Catalog # ABNOPAB18671
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitously-expressed Transcript
- Antigen symbol:UXT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Format:Liquid
- Gene ID:8409
- Antigen synonyms:ART-27, STAP1
- Storage buffer:Tris saline, pH 7,3 (0,02% sodium azide, 0,5% BSA)
- Sequence:C-DHRDKVYEQLAKY
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids at C-terminus of human UXT.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of UXT.
Recommended Dilutions: ELISA: 1:8000; Western Blot: 1-3 ?g/ml
Type: Primary
Antigen: UXT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Goat
Isotype:
Reactivity: Human