Order Entry
Singapore
ContactUsLinkComponent
Anti-DPF1 Rabbit Polyclonal Antibody
Anti-DPF1 Rabbit Polyclonal Antibody
  ABNOPAB18794
 :  Abnova
Anti-DPF1 Rabbit Polyclonal Antibody
  ABNOPAB18794
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    D4, Zinc And double PHD Fingers family 1
  • Antigen symbol:
    DPF1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    8193
  • Antigen synonyms:
    neuro-d4|NEUD4|BAF45b
  • Storage buffer:
    buffer containing 0,02% sodium azide
  • Storage temperature:
    Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to amino acids at C-terminus of human DPF1.
  • Purification:
    Protein A purification
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Rabbit polyclonal antibody raised against synthetic peptide of DPF1.

Recommended Dilutions: ELISA: 1:1000-1:10000; Western Blot: 1:200-1:1000

Type: Primary
Antigen: DPF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat