Anti-HK3 Rabbit Polyclonal Antibody
Catalog # ABNOPAB17875
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Hexokinase 3
- Antigen symbol:HK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:3101
- Antigen synonyms:HXK3|HKIII
- Storage buffer:PBS (0,09% sodium azide)
- Storage temperature:Store at –20 °C. For long term storage store at –80 °C
- Immunogen:A synthetic peptide corresponding to C-terminus of human HK3.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of HK3.
Recommended Dilutions: ELISA: 0.25 ?g/ml
Type: Primary
Antigen: HK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human